Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AACACACCUGGUUAACCUUUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-329a | MIMAT0009288 |
| Sheep | oar-miR-329a-3p | MIMAT0019266 |
Stem-loop Details
Bovine
Sheep

Genomic Map