Assay Details
Mature miRNA Sequence: | CCAAUUUUUAUCAAGGAAAGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dictyostelium discoideum | ddi-miR-1176 | MIMAT0005531 |
Stem-loop Details
Dictyostelium discoideum
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ddi-miR-1176 |
|---|---|
| miRBase Accession Number: | MI0006244 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCAAUUUUUAUCAAGGAAAGC |
| Species: | Dictyostelium discoideum |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CCAAUUUUUAUCAAGGAAAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dictyostelium discoideum | ddi-miR-1176 | MIMAT0005531 |