Assay Details
Mature miRNA Sequence: | UGCAGUCCAUGGGCAUAUACAC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-455-3p | MIMAT0006788 |
Columba livia | cli-miR-455-3p | MIMAT0038647 |
Stem-loop Details
Chicken
Columba livia
