Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACACUUCCCGCCCUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila erecta | der-miR-310a-3p | MIMAT0008553 |
| Drosophila yakuba | dya-miR-310-3p | MIMAT0009092 |
Stem-loop Details
Drosophila erecta
Drosophila yakuba
More Information
Related Products
miRBase Alias:
Drosophila erecta: der-miR-310a (v21)
Drosophila yakuba: dya-miR-310 (v21)
Drosophila yakuba: dya-miR-310 (v21)

Genomic Map