Assay Details
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | CCUCGUAGCUUGAUCACGAUAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-1329-3p | MIMAT0007282 |
Stem-loop Details
Chicken
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | gga-miR-1329* |
|---|---|
| miRBase Accession Number: | MI0006979 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUCGUAGCUUGAUCACGAUAU |
| Species: | Chicken |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | CCUCGUAGCUUGAUCACGAUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-1329-3p | MIMAT0007282 |