Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | GUGACAUCACAUAUAUGGCGAC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-489 | MIMAT0009942 |
Stem-loop Details
Bovine

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bta-miR-489 |
---|---|
miRBase Accession Number: | MI0010441 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GUGACAUCACAUAUAUGGCGAC |
Species: | Bovine |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | GUGACAUCACAUAUAUGGCGAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-489 | MIMAT0009942 |