Assay Details
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | UCACAACCUCCUUGAGUGAGCGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-307a-3p | MIMAT0000398 |
| Bactrocera dorsalis | bdo-miR-307 | MIMAT0045374 |
| Locusta migratoria | lmi-miR-307-3p | MIMAT0010137 |
| Manduca sexta | mse-miR-307 | MIMAT0024448 |
Stem-loop Details
Drosophila melanogaster
Bactrocera dorsalis
Locusta migratoria
Manduca sexta

Genomic Map