Assay Details
Mature miRNA Sequence: | AACCCGUAGUCAAGUCUACGGAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2274 | MIMAT0011733 |
Stem-loop Details
Pristionchus pacificus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppc-miR-2274 |
|---|---|
| miRBase Accession Number: | MI0011241 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACCCGUAGUCAAGUCUACGGAG |
| Species: | Pristionchus pacificus |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | AACCCGUAGUCAAGUCUACGGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2274 | MIMAT0011733 |