Assay Details
Mature miRNA Sequence: | UUUUCAUAACAGCUCUGUCAGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2791 | MIMAT0013689 |
Stem-loop Details
Bombyx mori

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bmo-miR-2791 |
---|---|
miRBase Accession Number: | MI0012364 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUUUCAUAACAGCUCUGUCAGC |
Species: | Bombyx mori |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUUUCAUAACAGCUCUGUCAGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2791 | MIMAT0013689 |