Assay Details
Mature miRNA Sequence: | UAUACGGUUCGGAAUCCUCUAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2563-3p | MIMAT0012389 |
Stem-loop Details
Drosophila pseudoobscura
More Information
Related Products
miRBase Alias:
Derxomyces pseudoschimicola: dps-miR-2563* (v18)

Genomic Map