Assay Details
Gene Family ID: | MIPF0001951, mir-2331 |
Mature miRNA Sequence: | GGCUUCCGUGCCUGCAGAUGUCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2331-5p | MIMAT0011863 |
Stem-loop Details
Bovine

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bta-miR-2331 |
---|---|
miRBase Accession Number: | MI0011357 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GGCUUCCGUGCCUGCAGAUGUCU |
Species: | Bovine |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001951, mir-2331 |
Mature miRNA Sequence: | GGCUUCCGUGCCUGCAGAUGUCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2331-5p | MIMAT0011863 |