Assay Details
Gene Family ID: | MIPF0000829, mir-2774 |
Mature miRNA Sequence: | CUCGGUGGAAGAUCGUCCCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2774a | MIMAT0013658 |
Stem-loop Details
Bombyx mori

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bmo-miR-2774a |
---|---|
miRBase Accession Number: | MI0012331 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CUCGGUGGAAGAUCGUCCCU |
Species: | Bombyx mori |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000829, mir-2774 |
Mature miRNA Sequence: | CUCGGUGGAAGAUCGUCCCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2774a | MIMAT0013658 |