Assay Details
Mature miRNA Sequence: | AGGCAGAUCAAUCAAUUUUUAGG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis elegans | cel-miR-2210-5p | MIMAT0011435 |
Stem-loop Details
Caenorhabditis elegans

More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2210 (v17)
