Assay Details
Gene Family ID: | MIPF0001013, mir-303 |
Mature miRNA Sequence: | UUGGCAUUAAAGGAAGCUAGUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila simulans | dsi-miR-303-5p | MIMAT0012516 |
Stem-loop Details
Drosophila simulans
