Assay Details
Mature miRNA Sequence: | AGCUCGUGUCCCAAGGCGCCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Zebrafish | dre-miR-2198 | MIMAT0011305 |
Stem-loop Details
Zebrafish
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dre-miR-2198 |
|---|---|
| miRBase Accession Number: | MI0010847 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCUCGUGUCCCAAGGCGCCU |
| Species: | Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | AGCUCGUGUCCCAAGGCGCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Zebrafish | dre-miR-2198 | MIMAT0011305 |