More Information
Updated Mapping Information
miRBase Alias:
Caenorhabditis elegans: cel-miR-2214 (v17)
Caenorhabditis elegans: cel-miR-2214-3p (v20)
Caenorhabditis elegans: cel-miR-2214-3p (v20)
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cel-miR-2214 |
|---|---|
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAUUGACAACAACUUGACCGGCGUU |
| Product Type: | TaqMan™ MicroRNA Assay |