Assay Details
Gene Family ID: | MIPF0000977, mir-2511 |
Mature miRNA Sequence: | UAAGUCUGUUGAGUGCCAUAGA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila pseudoobscura | dps-miR-2511 | MIMAT0012296 |
Stem-loop Details
Drosophila pseudoobscura
