Assay Details
Gene Family ID: | MIPF0000134, mir-460 |
Mature miRNA Sequence: | CCCUCAUUGUACAUGCUGUGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-460b-5p | MIMAT0021972 |
| Taeniopygia guttata | tgu-miR-460b-5p | MIMAT0014476 |
Stem-loop Details
Anolis carolinensis
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Trabulsiella guamensis ATCC 49490: tgu-miR-460b (v18)

Genomic Map