Assay Details
Gene Family ID: | MIPF0000975, mir-2494 |
Mature miRNA Sequence: | UUCCCAGUAAUGUCCACUUACAAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-2494-3p | MIMAT0012203 |
Stem-loop Details
Drosophila melanogaster
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-2494 (v16)

Genomic Map