Assay Details
Mature miRNA Sequence: | ACGCACUGUCCUGGGAAGUGG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2456 | MIMAT0012041 |
Stem-loop Details
Bovine

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bta-miR-2456 |
---|---|
miRBase Accession Number: | MI0011515 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACGCACUGUCCUGGGAAGUGG |
Species: | Bovine |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | ACGCACUGUCCUGGGAAGUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2456 | MIMAT0012041 |