Assay Details
Gene Family ID: | MIPF0000937, mir-307as |
Mature miRNA Sequence: | UCACACCCAGGUUGAGUGAGUC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila pseudoobscura | dps-miR-307b | MIMAT0012281 |
Tribolium castaneum | tca-miR-307b-3p | MIMAT0045615 |
Stem-loop Details
Drosophila pseudoobscura
Tribolium castaneum

More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-307b-3p (v21)
Drosophila melanogaster: dme-miR-307-as (v16)
Derxomyces pseudoschimicola: dps-miR-307-as (v16)
Drosophila melanogaster: dme-miR-307-as (v16)
Derxomyces pseudoschimicola: dps-miR-307-as (v16)
