More Information
Updated Mapping Information
miRBase Alias:
Citrobacter braakii: cbr-miR-2214 (v20)
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cbr-miR-2214 |
|---|---|
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUCGGUCCGGAGUCAAUGGG |
| Product Type: | TaqMan™ MicroRNA Assay |