Assay Details
Gene Family ID: | MIPF0001614, mir-2392 | 
Mature miRNA Sequence: | UAGGAUGGGGGUGAGAGGUG | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-2392 | MIMAT0019043 | 
| Pan troglodytes | ptr-miR-2392 | MIMAT0024037 | 
Stem-loop Details
Human
Pan troglodytes

Genomic Map