Assay Details
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | ACUCACUCAACCUGGGUGUGAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-307a-5p | MIMAT0020832 |
| Dinoponera quadriceps | dqu-miR-307-5p | MIMAT0045733 |
Stem-loop Details
Drosophila melanogaster
Dinoponera quadriceps

Genomic Map