Assay Details
Gene Family ID: | MIPF0000163, mir-320 | 
Mature miRNA Sequence: | GCCUUCUCUUCCCGGUUCUUCC | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-320a-5p | MIMAT0037311 | 
| Mouse | mmu-miR-320-5p | MIMAT0017057 | 
| Rat | rno-miR-320-5p | MIMAT0017168 | 
| Pteropus alecto | pal-miR-320-5p | MIMAT0040073 | 
Stem-loop Details
Human
Mouse
Rat
Pteropus alecto
More Information
Related Products
miRBase Alias:
						
							Mouse: mmu-miR-320* (v17)
Rat: rno-miR-320* (v18)
						
					
				Rat: rno-miR-320* (v18)

Genomic Map