Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 15 - 52134129 - 52134129 [+] on 52134129 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-320-3p | MIMAT0000903 |
| Rat | rno-miR-320-5p | MIMAT0017168 |
Stem-loop Details
Rat

Genomic Map