Assay Details
Amplicon Length: | 55 |
Chromosome Location: | Chr. 8 - 22244564 - 22244564 [-] on 22244564 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-320a-5p | MIMAT0037311 |
| Human | hsa-miR-320a-3p | MIMAT0000510 |
Stem-loop Details
Human

Genomic Map