Assay Details
Amplicon Length: | 99 |
Chromosome Location: | Chr. 14 - 70443305 - 70443305 [+] on 70443305 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-320-3p | MIMAT0000666 |
| Mouse | mmu-miR-320-5p | MIMAT0017057 |
Stem-loop Details
Mouse

Genomic Map