Assay Details
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GCGACCCAUACUUGGUUUCAAA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Petromyzon marinus | pma-miR-551 | MIMAT0019558 |
Stem-loop Details
Petromyzon marinus
