Assay Details
Gene Family ID: | MIPF0000046, mir-101 |
Mature miRNA Sequence: | UCGGUUAUCAUGGUACCGAUGCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-101b-5p | MIMAT0017046 |
Dasypus novemcinctus | dno-miR-101-2-5p | MIMAT0047600 |
Guinea pig | cpo-miR-101-2-5p | MIMAT0046949 |
Monodelphis domestica | mdo-miR-101-2-5p | MIMAT0026657 |
Rabbit | ocu-miR-101-2-5p | MIMAT0048188 |
Rhesus monkey | mml-miR-101-2-5p | MIMAT0031100 |
Stem-loop Details
Mouse
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Rabbit
Rhesus monkey
