Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | GAAGGUGCUUCCCUUUGCAGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rhesus monkey | mml-miR-524-3p | MIMAT0026950 |
Stem-loop Details
Rhesus monkey
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mml-miR-524-3p |
|---|---|
| miRBase Accession Number: | MI0012534 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAAGGUGCUUCCCUUUGCAGUG |
| Species: | Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | GAAGGUGCUUCCCUUUGCAGUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rhesus monkey | mml-miR-524-3p | MIMAT0026950 |