Assay Details
Mature miRNA Sequence: | CAAGGAUCCCUGACUUGAUCAU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Monodelphis domestica | mdo-miR-1547-3p | MIMAT0026771 |
Stem-loop Details
Monodelphis domestica

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mdo-miR-1547-3p |
---|---|
miRBase Accession Number: | MI0007272 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAAGGAUCCCUGACUUGAUCAU |
Species: | Monodelphis domestica |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CAAGGAUCCCUGACUUGAUCAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Monodelphis domestica | mdo-miR-1547-3p | MIMAT0026771 |