Assay Details
Gene Family ID: | MIPF0001854, mir-2067 |
Mature miRNA Sequence: | ACUGUUGCAUCUUGCUGCCUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2067-5p | MIMAT0031631 |
Stem-loop Details
Branchiostoma belcheri

Genomic Map