Assay Details
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | CCUCGUAGCUUGAUCACGAUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Taeniopygia guttata | tgu-miR-1329-3p | MIMAT0026956 |
Stem-loop Details
Taeniopygia guttata
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | tgu-miR-1329-3p |
|---|---|
| miRBase Accession Number: | MI0013635 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUCGUAGCUUGAUCACGAUA |
| Species: | Taeniopygia guttata |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | CCUCGUAGCUUGAUCACGAUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Taeniopygia guttata | tgu-miR-1329-3p | MIMAT0026956 |