Assay Details
| Gene Family ID: | MIPF0000110, mir-329 | 
| Mature miRNA Sequence: | GAGGUUUUCUGGGUCUCUGUUU | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Rhesus monkey | mml-miR-329-2-5p | MIMAT0031107 | 
Stem-loop Details
Rhesus monkey
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top mirVana® miRNA inhibitor :
						mirVana® miRNA inhibitor :