Assay Details
Gene Family ID: | MIPF0001793, mir-2058 |
Mature miRNA Sequence: | UGAGAAGUAAGACUACCAUCCCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2058-5p | MIMAT0031615 |
Stem-loop Details
Branchiostoma belcheri

Genomic Map