Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAACAUGGUCCACUAAC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dasypus novemcinctus | dno-miR-379-3p | MIMAT0047861 |
Goat | chi-miR-379-3p | MIMAT0036196 |
Guinea pig | cpo-miR-379-3p | MIMAT0047222 |
Pteropus alecto | pal-miR-379-3p | MIMAT0039887 |
Rabbit | ocu-miR-379-3p | MIMAT0048424 |
Rhesus monkey | mml-miR-379-3p | MIMAT0026865 |
Stem-loop Details
Dasypus novemcinctus
Goat
Guinea pig
Pteropus alecto
Rabbit
Rhesus monkey
