Assay Details
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GCGACCCACUCUUGGUUUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-551a | MIMAT0025379 |
Stem-loop Details
Pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ssc-miR-551a |
|---|---|
| miRBase Accession Number: | MI0022153 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCGACCCACUCUUGGUUUCC |
| Species: | Pig |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GCGACCCACUCUUGGUUUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-551a | MIMAT0025379 |