Assay Details
Mature miRNA Sequence: | UGAUAUGUUUGAUAUAUUAGGUUG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Astatotilapia burtoni | abu-miR-190a-5p | MIMAT0042144 | |
Cricetulus griseus | cgr-miR-190a | MIMAT0023819 | |
Dasypus novemcinctus | dno-miR-190a-5p | MIMAT0047713 | |
Guinea pig | cpo-miR-190a-5p | MIMAT0047062 | |
Metriaclima zebra | mze-miR-190a | MIMAT0042379 | |
Neolamprologus brichardi | nbr-miR-190a | MIMAT0042554 | |
View More Rows | |||
Nomascus leucogenys | nle-miR-190 | MIMAT0049423 | |
Oreochromis niloticus | oni-miR-190a | MIMAT0042741 | |
Otolemur garnettii | oga-miR-190 | MIMAT0049719 | |
Pundamilia nyererei | pny-miR-190a | MIMAT0042941 | |
Python bivittatus | pbv-miR-190a-5p | MIMAT0038984 | |
Rabbit | ocu-miR-190a-5p | MIMAT0048299 | |
Salmo salar | ssa-miR-190a-5p | MIMAT0032436 |
Stem-loop Details
Astatotilapia burtoni
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Metriaclima zebra
Neolamprologus brichardi
Nomascus leucogenys
Oreochromis niloticus
Otolemur garnettii
Pundamilia nyererei
Python bivittatus
Rabbit
Salmo salar
