Assay Details
Mature miRNA Sequence: | CUGAACUGUCUUACUCCCACAUC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Goat | chi-miR-2411-3p | MIMAT0036094 |
Stem-loop Details
Goat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | chi-miR-2411-3p |
---|---|
miRBase Accession Number: | MI0030715 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CUGAACUGUCUUACUCCCACAUC |
Species: | Goat |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CUGAACUGUCUUACUCCCACAUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Goat | chi-miR-2411-3p | MIMAT0036094 |