Assay Details
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | CUCCCACAUGCAAGGUUUGCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tupaia chinensis | tch-miR-188-3p | MIMAT0036492 |
Stem-loop Details
Tupaia chinensis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | tch-miR-188-3p |
|---|---|
| miRBase Accession Number: | MI0031149 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCCCACAUGCAAGGUUUGCA |
| Species: | Tupaia chinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | CUCCCACAUGCAAGGUUUGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tupaia chinensis | tch-miR-188-3p | MIMAT0036492 |