Assay Details
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | CCCAAAGGUGAAUUUUUUGG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Goat | chi-miR-186-3p | MIMAT0036011 |
Stem-loop Details
Goat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | chi-miR-186-3p |
---|---|
miRBase Accession Number: | MI0030657 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CCCAAAGGUGAAUUUUUUGG |
Species: | Goat |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | CCCAAAGGUGAAUUUUUUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Goat | chi-miR-186-3p | MIMAT0036011 |