Assay Details
Gene Family ID: | MIPF0001630, mir-4428 |
Mature miRNA Sequence: | CAAGGAGACGGGAACAUGGAGCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pan troglodytes | ptr-miR-4428 | MIMAT0024069 |
Stem-loop Details
Pan troglodytes
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ptr-miR-4428 |
|---|---|
| miRBase Accession Number: | MI0020616 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAGGAGACGGGAACAUGGAGCC |
| Species: | Pan troglodytes |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001630, mir-4428 |
Mature miRNA Sequence: | CAAGGAGACGGGAACAUGGAGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pan troglodytes | ptr-miR-4428 | MIMAT0024069 |