Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | GAACAUCACAGCAAGUCUGUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-499-3p | MIMAT0023973 |
| Goat | chi-miR-499-3p | MIMAT0036262 |
Stem-loop Details
Cricetulus griseus
Goat

Genomic Map