Assay Details
Gene Family ID: | MIPF0000134, mir-460 |
Mature miRNA Sequence: | CACAGCGCAUACAAUGUGGAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-460a-3p | MIMAT0021971 |
| Chrysemys picta | cpi-miR-460a-3p | MIMAT0037895 |
| Gadus morhua | gmo-miR-460-3p | MIMAT0044249 |
Stem-loop Details
Anolis carolinensis
Chrysemys picta
Gadus morhua

Genomic Map