Assay Details
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | CCAGUAUUGCCUGUGCUGCUCCA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Anolis carolinensis | aca-miR-457-3p | MIMAT0021968 |
Stem-loop Details
Anolis carolinensis
