Assay Details
Gene Family ID: | MIPF0000812, mir-2188 |
Mature miRNA Sequence: | AAGGUCCAACCUCAUGCGUCCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ophiophagus hannah | oha-miR-2188-5p | MIMAT0036839 |
Stem-loop Details
Ophiophagus hannah
