Assay Details
Gene Family ID: | MIPF0000137, mir-383 |
Mature miRNA Sequence: | CACAGCACUGCCUGGUCAGAA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Anolis carolinensis | aca-miR-383-3p | MIMAT0021953 |
Stem-loop Details
Anolis carolinensis
