Assay Details
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | CAGUGCAAUAGUAUUGUCAAAGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-301a-3p | MIMAT0000688 |
| Mouse | mmu-miR-301a-3p | MIMAT0000379 |
| Rat | rno-miR-301a-3p | MIMAT0000552 |
| Anolis carolinensis | aca-miR-301a-3p | MIMAT0021916 |
| Callithrix jacchus | cja-miR-301a | MIMAT0049307 |
| Chrysemys picta | cpi-miR-301a-3p | MIMAT0037858 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-301a-3p | MIMAT0023891 |
| Cyprinus carpio | ccr-miR-301a | MIMAT0026288 |
| Dog | cfa-miR-301a | MIMAT0009853 |
| Goat | chi-miR-301a-3p | MIMAT0036119 |
| Gorilla gorilla | ggo-miR-301a | MIMAT0049111 |
| Horse | eca-miR-301a | MIMAT0013032 |
| Ictalurus punctatus | ipu-miR-301a | MIMAT0029494 |
| Microcebus murinus | mmr-miR-301a | MIMAT0049153 |
| Monodelphis domestica | mdo-miR-301-3p | MIMAT0012739 |
| Nomascus leucogenys | nle-miR-301a | MIMAT0049397 |
| Ornithorhynchus anatinus | oan-miR-301-3p | MIMAT0006910 |
| Otolemur garnettii | oga-miR-301a | MIMAT0049593 |
| Pan troglodytes | ptr-miR-301a | MIMAT0008084 |
| Pongo pygmaeus | ppy-miR-301a | MIMAT0049335 |
| Python bivittatus | pbv-miR-301a-3p | MIMAT0039039 |
| Rhesus monkey | mml-miR-301a-3p | MIMAT0006257 |
| Salmo salar | ssa-miR-301d-3p | MIMAT0032599 |
| Taeniopygia guttata | tgu-miR-301-3p | MIMAT0014595 |
| Xenopus tropicalis | xtr-miR-301 | MIMAT0003635 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-301a |
| Stem-loop Accession # | MI0000745 |
| Stem-loop Sequence | ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGG |
| Chromosome Location | Chr. 17 - 59151136 - 59151221 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000688
miRBase ID: hsa-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: Chr. 17 - 59151136 - 59151221 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-301a |
| Stem-loop Accession # | MI0000401 |
| Stem-loop Sequence | CCUGCUAACGGCUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCGAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGCGAGCAGG |
| Chromosome Location | Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000379
miRBase ID: mmu-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-301a |
| Stem-loop Accession # | MI0000593 |
| Stem-loop Sequence | CCUGCUGGCUACUGCUGACGACUGCUCUGACUUUAUUGCACUACUGUACUGUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGGGAGCAGGCUAC |
| Chromosome Location | Chr. 10 - 74417746 - 74417845 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000552
miRBase ID: rno-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: Chr. 10 - 74417746 - 74417845 [+] on Build Rnor_6.0 |
| Stem-loop ID | aca-mir-301a |
| Stem-loop Accession # | MI0018844 |
| Stem-loop Sequence | UUCCUGCUGCGGACAAGGGCUCUGACUUCAUUGCACUACUGUACUUCCCAACUAGCAGUGCAAUAGUAUUGUCAAAGCACUUGCAAGCAGAACUGACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0021916
miRBase ID: aca-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | cja-mir-301a |
| Stem-loop Accession # | MI0040043 |
| Stem-loop Sequence | UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049307
miRBase ID: cja-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | cpi-mir-301a |
| Stem-loop Accession # | MI0029550 |
| Stem-loop Sequence | UGCUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037858
miRBase ID: cpi-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | cgr-mir-301a |
| Stem-loop Accession # | MI0020482 |
| Stem-loop Sequence | GCUCGCGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGAAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023891
miRBase ID: cgr-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | ccr-mir-301a |
| Stem-loop Accession # | MI0023388 |
| Stem-loop Sequence | ACAUCAAGUGCUGUUAACAGGUGCUCUGACUUCAUUGCACUACUGUAUUGGACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAGAGCAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026288
miRBase ID: ccr-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | cfa-mir-301a |
| Stem-loop Accession # | MI0010348 |
| Stem-loop Sequence | CUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009853
miRBase ID: cfa-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | chi-mir-301a |
| Stem-loop Accession # | MI0030729 |
| Stem-loop Sequence | ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036119
miRBase ID: chi-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | ggo-mir-301a |
| Stem-loop Accession # | MI0039823 |
| Stem-loop Sequence | UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049111
miRBase ID: ggo-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | eca-mir-301a |
| Stem-loop Accession # | MI0012779 |
| Stem-loop Sequence | ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013032
miRBase ID: eca-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | ipu-mir-301a |
| Stem-loop Accession # | MI0024611 |
| Stem-loop Sequence | UCUGACUUCAUUGCACUACUGUAUUGGACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0029494
miRBase ID: ipu-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | mmr-mir-301a |
| Stem-loop Accession # | MI0039868 |
| Stem-loop Sequence | UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049153
miRBase ID: mmr-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | mdo-mir-301 |
| Stem-loop Accession # | MI0012495 |
| Stem-loop Sequence | ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012739
miRBase ID: mdo-miR-301-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | nle-mir-301a |
| Stem-loop Accession # | MI0040137 |
| Stem-loop Sequence | UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049397
miRBase ID: nle-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | oan-mir-301 |
| Stem-loop Accession # | MI0006727 |
| Stem-loop Sequence | UAUUAUGGUGUGUUUUACGCUUUCUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUUCUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGAAUACACAGCAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006910
miRBase ID: oan-miR-301-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | oga-mir-301a |
| Stem-loop Accession # | MI0040374 |
| Stem-loop Sequence | UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049593
miRBase ID: oga-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | ptr-mir-301a |
| Stem-loop Accession # | MI0008597 |
| Stem-loop Sequence | ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008084
miRBase ID: ptr-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | ppy-mir-301a |
| Stem-loop Accession # | MI0040071 |
| Stem-loop Sequence | UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049335
miRBase ID: ppy-miR-301a Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | pbv-mir-301a |
| Stem-loop Accession # | MI0030300 |
| Stem-loop Sequence | UUUUGCUGCUGACAAGUGCUCUGACUUCAUUGCACUACUGUACUUCCCAGCUAGCAGUGCAAUAGUAUUGUCAAAGCACUUGCGAGCAGAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039039
miRBase ID: pbv-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | mml-mir-301a |
| Stem-loop Accession # | MI0007685 |
| Stem-loop Sequence | ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006257
miRBase ID: mml-miR-301a-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | ssa-mir-301d |
| Stem-loop Accession # | MI0026673 |
| Stem-loop Sequence | CUCUGACUUCAUUGCACUACUGUAUUGGACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032599
miRBase ID: ssa-miR-301d-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | tgu-mir-301 |
| Stem-loop Accession # | MI0013823 |
| Stem-loop Sequence | AGUGCUCUGACUUUAUUGCACUACUGUACUAUACAGAGAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAGAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0014595
miRBase ID: tgu-miR-301-3p Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | xtr-mir-301-1 |
| Stem-loop Accession # | MI0004876 |
| Stem-loop Sequence | CUGCUAAUGUGUGCUCUGACUUUAUUGCACUACUGUACUAUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGCAAGCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003635
miRBase ID: xtr-miR-301 Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
| Stem-loop ID | xtr-mir-301-2 |
| Stem-loop Accession # | MI0004877 |
| Stem-loop Sequence | GAAUUGCUCUGACAGUGUUGCACUACUGUGAUCUCAAAUGAAGCAGUGCAAUAGUAUUGUCAAAGCAUUUCAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003635
miRBase ID: xtr-miR-301 Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC Chromosome Location: NA |
More Information
Related Products

Genomic Map