Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CCGCACUGUGGGUACUUGCUGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-106b-3p | MIMAT0004672 |
Mouse | mmu-miR-106b-3p | MIMAT0004582 |
Rat | rno-miR-106b-3p | MIMAT0004727 |
Cricetulus griseus | cgr-miR-106b-3p | MIMAT0023733 |
Pteropus alecto | pal-miR-106b-3p | MIMAT0039936 |
Stem-loop Details
Human
Mouse
Rat
Cricetulus griseus
Pteropus alecto
